(R)-(+)-tert-Butylsulfinamide, 98%
(R)-(+)-tert-Butylsulfinamide, 98%
(R)-(+)-tert-Butylsulfinamide, 98%
Thermo Scientific Chemicals

(R)-(+)-tert-Butylsulfinamide, 98%

CAS: 196929-78-9 | C4H11NOS | 121.20 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
產品號碼 391100050
價格 (TWD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS196929-78-9
IUPAC Name2-methylpropane-2-sulfinamide
Molecular FormulaC4H11NOS
InChI KeyCESUXLKAADQNTB-UHFFFAOYNA-N
SMILESCC(C)(C)S(N)=O
檢視更多
規格Specification Sheet規格表
GC>=97.5 %
Infrared spectrumConforms
Specific optical rotation3.0° to 5.0° (20°C, 589 nm) (c=1, chloroform)
Appearance (Form)Crystalline powder
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only