4-Diphenylaminobenzaldehyde, 98%, Thermo Scientific Chemicals
4-Diphenylaminobenzaldehyde, 98%, Thermo Scientific Chemicals
4-Diphenylaminobenzaldehyde, 98%, Thermo Scientific Chemicals

4-Diphenylaminobenzaldehyde, 98%, Thermo Scientific Chemicals

CAS: 4181-05-9 | Molecular Formula: C19H15NO | Molecular Weight: 273.33 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3911800505 gGlass bottle
Catalog number 391180050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4181-05-9
IUPAC Name4-(diphenylamino)benzaldehyde
Molecular FormulaC19H15NO
InChI KeyUESSERYYFWCTBU-UHFFFAOYSA-N
SMILESO=CC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or chunks
Appearance (Color)White to yellow to brown
Water=<2 % (K.F.)
HPLC>=97.5 %
Infrared spectrumConforms
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only