1,4-Diethynylbenzene, 95%
1,4-Diethynylbenzene, 95%
1,4-Diethynylbenzene, 95%
Thermo Scientific Chemicals

1,4-Diethynylbenzene, 95%

CAS: 935-14-8 | C10H6 | 126.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 391550010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS935-14-8
IUPAC Name1,4-diethynylbenzene
Molecular FormulaC10H6
InChI KeyMVLGANVFCMOJHR-UHFFFAOYSA-N
SMILESC#CC1=CC=C(C=C1)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder, crystals and/or chunks
GC>=94 %
Infrared spectrumConforms
Appearance (Color)White to orange to brown
Melting point90°C to 96°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only