4-Biphenylsulfonyl chloride, 97%
4-Biphenylsulfonyl chloride, 97%
4-Biphenylsulfonyl chloride, 97%
Thermo Scientific Chemicals

4-Biphenylsulfonyl chloride, 97%

CAS: 1623-93-4 | C12H9ClO2S | 252.71 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3919600505 gGlass bottle
Catalog number 391960050
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
62.50
Save 8.85
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
62.50
Save 8.85
Each
Chemical Identifiers
CAS1623-93-4
IUPAC Name[1,1'-biphenyl]-4-sulfonyl chloride
Molecular FormulaC12H9ClO2S
InChI KeyALBQXDHCMLLQMB-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Appearance (Color)White to cream
Infrared spectrumConforms
Appearance (Form)Powder
Melting point111°C to 116°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only