Dimethylethoxysilane, 94%
Dimethylethoxysilane, 94%
Dimethylethoxysilane, 94%
Thermo Scientific Chemicals

Dimethylethoxysilane, 94%

CAS: 14857-34-2 | C4H12OSi | 104.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 392001000
Price (USD)
208.65
Special offer
Online exclusive
Ends: 30-Jun-2026
245.00
Save 36.35 (15%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
208.65
Special offer
Online exclusive
Ends: 30-Jun-2026
245.00
Save 36.35 (15%)
Each
Chemical Identifiers
CAS14857-34-2
IUPAC Nameethoxydimethylsilyl
Molecular FormulaC4H11OSi
InChI KeyDRUOQOFQRYFQGB-UHFFFAOYSA-N
SMILESCCO[Si](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
Refractive index1.3670 to 1.3700 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=92 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only