3-Iodobenzaldehyde, 99%
3-Iodobenzaldehyde, 99%
3-Iodobenzaldehyde, 99%
Thermo Scientific Chemicals

3-Iodobenzaldehyde, 99%

CAS: 696-41-3 | C7H5IO | 232.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 392070010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS696-41-3
IUPAC Name3-iodobenzaldehyde
Molecular FormulaC7H5IO
InChI KeyRZODAQZAFOBFLS-UHFFFAOYSA-N
SMILESIC1=CC=CC(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Melting point55°C to 59°C
Appearance (Form)Crystalline powder or crystals or needles
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only