2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized
2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized
2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized
Thermo Scientific Chemicals

2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized

2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized, Quantity: 2.5L, Packaging: Glass bottle, Boiling Point: 78.0 deg.C to 80.0 deg.C, CAS: 96-47-9, Colorless, Grade: For Grignard reaction, Melting Point: -136.0 deg.C, Molecular Formula: C5H10O | CAS: 96-47-9 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Metal Drum
Catalog number 392380100
Price (USD)
809.65
Special offer
Online exclusive
Ends: 28-Dec-2026
952.00
Save 142.35
Each
Quantity:
10 L
Packaging:
Metal Drum
Request bulk or custom format
Price (USD)
809.65
Special offer
Online exclusive
Ends: 28-Dec-2026
952.00
Save 142.35
Each
Chemical Identifiers
CAS96-47-9
IUPAC Name2-methyloxolane
Molecular FormulaC5H10O
InChI KeyJWUJQDFVADABEY-UHFFFAOYNA-N
SMILESCC1CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=99.0 %
Stabilizer150 to 400 ppm BHT
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only