Ethyl 4-bromophenylacetate, 99%
Ethyl 4-bromophenylacetate, 99%
Ethyl 4-bromophenylacetate, 99%
Thermo Scientific Chemicals

Ethyl 4-bromophenylacetate, 99%

CAS: 14062-25-0 | C10H11BrO2 | 243.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 392990250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14062-25-0
IUPAC Nameethyl 2-(4-bromophenyl)acetate
Molecular FormulaC10H11BrO2
InChI KeyZFDCWHPNBWPPHG-UHFFFAOYSA-N
SMILESCCOC(=O)CC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid or low melting solid
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only