Ammonium bicarbonate, 99%, for analysis
Ammonium bicarbonate, 99%, for analysis
Ammonium bicarbonate, 99%, for analysis
Thermo Scientific Chemicals

Ammonium bicarbonate, 99%, for analysis

CAS: 1066-33-7 | C3H8N2O9Zn2 | 346.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 393212500
Price (USD)
46.65
Online exclusive
51.70
Save 5.05
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
46.65
Online exclusive
51.70
Save 5.05
Each
Chemical Identifiers
CAS1066-33-7
IUPAC Namecarbonic acid amine
Molecular FormulaCH5NO3
InChI KeyATRRKUHOCOJYRX-UHFFFAOYSA-N
SMILESN.OC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<5 ppm
Potassium (K)=<10 ppm
Sodium (Na)=<20 ppm
Heavy metals (as Pb)=<5 ppm
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only