Ácido bencilfosfónico, 98 %, Thermo Scientific Chemicals
Ácido bencilfosfónico, 98 %, Thermo Scientific Chemicals
Ácido bencilfosfónico, 98 %, Thermo Scientific Chemicals
Ácido bencilfosfónico, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ácido bencilfosfónico, 98 %, Thermo Scientific Chemicals

CAS: 6881-57-8 | C7H7O3P | 170.11 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
25 g
Envase:
Frasco de vidrio
Número de catálogo 393340010
Precio (USD)
-
Cantidad:
1 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS6881-57-8
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
Melting point169°C to 173°C
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only