(S)-4-(1-Hydroxyethyl)pyridine, 99+%, (99+% ee)
(S)-4-(1-Hydroxyethyl)pyridine, 99+%, (99+% ee)
(S)-4-(1-Hydroxyethyl)pyridine, 99+%, (99+% ee)
Thermo Scientific Chemicals

(S)-4-(1-Hydroxyethyl)pyridine, 99+%, (99+% ee)

CAS: 54656-96-1 | C7H9NO | 123.155 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number 393372500
Price (USD)
-
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS54656-96-1
IUPAC Name1-(pyridin-4-yl)ethan-1-ol
Molecular FormulaC7H9NO
InChI KeyHVOAMIOKNARIMR-UHFFFAOYNA-N
SMILESCC(O)C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or chunks
Appearance (Color)White to cream
Infrared spectrumConforms
Melting point67°C to 70°C
Specific optical rotation-56° to -61° (20°C, 589 nm) (c=1, chloroform)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only