4-Methoxypiperidine, 98+%
4-Methoxypiperidine, 98+%
4-Methoxypiperidine, 98+%
Thermo Scientific Chemicals

4-Methoxypiperidine, 98+%

CAS: 4045-24-3 | C6H13NO | 115.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 393390010
Price (USD)
131.65
Special offer
Online exclusive
Ends: 28-Dec-2026
155.00
Save 23.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
131.65
Special offer
Online exclusive
Ends: 28-Dec-2026
155.00
Save 23.35
Each
Chemical Identifiers
CAS4045-24-3
IUPAC Name4-methoxypiperidine
Molecular FormulaC6H13NO
InChI KeyZEYSHALLPAKUHG-UHFFFAOYSA-N
SMILESCOC1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=98.0 %
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4590 to 1.4610 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only