2-Ethoxycarbonylphenylboronic acid, 97%
2-Ethoxycarbonylphenylboronic acid, 97%
2-Ethoxycarbonylphenylboronic acid, 97%
Thermo Scientific Chemicals

2-Ethoxycarbonylphenylboronic acid, 97%

CAS: 380430-53-5 | C9H11BO4 | 193.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 393830250
Price (USD)
337.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
337.00
Each
Chemical Identifiers
CAS380430-53-5
IUPAC Name[2-(ethoxycarbonyl)phenyl]boronic acid
Molecular FormulaC9H11BO4
InChI KeyQZKVVOXAEBCLPZ-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=CC=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Solid
Infrared spectrumConforms
HPLC>=96.0 %
NMR (1H-NMR)Consistent with structure
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only