gamma-Hexalactone, 98%
gamma-Hexalactone, 98%
gamma-Hexalactone, 98%
Thermo Scientific Chemicals

gamma-Hexalactone, 98%

CAS: 695-06-7 | C6H10O2 | 114.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 394170250
Price (USD)
25.65
Special offer
Online exclusive
Ends: 28-Dec-2026
29.50
Save 3.85
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
25.65
Special offer
Online exclusive
Ends: 28-Dec-2026
29.50
Save 3.85
Each
Chemical Identifiers
CAS695-06-7
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4380 to 1.4400 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only