3-Hydroxyphthalic anhydride, 97%
3-Hydroxyphthalic anhydride, 97%
3-Hydroxyphthalic anhydride, 97%
Thermo Scientific Chemicals

3-Hydroxyphthalic anhydride, 97%

CAS: 37418-88-5 | C8H4O4 | 164.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 394350010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS37418-88-5
IUPAC Name4-hydroxy-1,3-dihydro-2-benzofuran-1,3-dione
Molecular FormulaC8H4O4
InChI KeyCCTOEAMRIIXGDJ-UHFFFAOYSA-N
SMILESOC1=CC=CC2=C1C(=O)OC2=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Appearance (Color)White to beige
Appearance (Form)Powder or crystals or flakes
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only