2-Chloroethyl chloroformate, 97%
2-Chloroethyl chloroformate, 97%
2-Chloroethyl chloroformate, 97%
Thermo Scientific Chemicals

2-Chloroethyl chloroformate, 97%

CAS: 627-11-2 | C3H4Cl2O2 | 142.96 g/mol
Have Questions?
Catalog NumberQuantityPackaging
395331000100 gGlass bottle
Catalog number 395331000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS627-11-2
IUPAC Name2-chloroethyl carbonochloridate
Molecular FormulaC3H4Cl2O2
InChI KeySVDDJQGVOFZBNX-UHFFFAOYSA-N
SMILESClCCOC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4450 to 1.4470 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only