Activated charcoal, NORIT™ GAC 1240, 12-40 mesh
Activated charcoal, NORIT™ GAC 1240, 12-40 mesh
Activated charcoal, NORIT™ GAC 1240, 12-40 mesh
Thermo Scientific Chemicals

Activated charcoal, NORIT™ GAC 1240, 12-40 mesh

CAS: 7440-44-0 | C | 12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 395712500
Price (USD)
72.65
Special offer
Online exclusive
Ends: 28-Dec-2026
85.80
Save 13.15
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
72.65
Special offer
Online exclusive
Ends: 28-Dec-2026
85.80
Save 13.15
Each
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Iodine number>=1000 mg/g
Molasses number>=230
Abrasion number>=75
Water=<5 % (As packed)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
RUO – Research Use Only