Sodium tetrachloroaurate hydrate, 98%
Sodium tetrachloroaurate hydrate, 98%
Sodium tetrachloroaurate hydrate, 98%
Thermo Scientific Chemicals

Sodium tetrachloroaurate hydrate, 98%

CAS: 29156-65-8 ∣ AuCl4Na ∣ 361.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 396270050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS29156-65-8
IUPAC Namesodium tetrachlorogolduide
Molecular FormulaAuCl4Na
InChI KeyIXPWAPCEBHEFOV-UHFFFAOYSA-J
SMILES[Na+].Cl[Au-](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals or granules
Assay48.3 to 50.3 % (Au)
Appearance (Color)Yellow to orange to golden
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only