1-Dodecanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
1-Dodecanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
1-Dodecanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
Thermo Scientific Chemicals

1-Dodecanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous

CAS: 2386-53-0 | C12H25NaO3S | 272.379 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 396720250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2386-53-0
IUPAC Namesodium dodecane-1-sulfonate
Molecular FormulaC12H25NaO3S
InChI KeyDAJSVUQLFFJUSX-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCS([O-])(=O)=O
さらに表示
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
UV(0.18 M in hot water)
UVat 220 nm A: =<0.03
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography
RUO – Research Use Only