Imidazole, ACS reagent
Imidazole, ACS reagent
Imidazole, ACS reagent
Thermo Scientific Chemicals

Imidazole, ACS reagent

CAS: 288-32-4 | C3H4N2 | 68.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 396740025
Price (USD)
716.65
Special offer
Online exclusive
Ends: 28-Dec-2026
843.00
Save 126.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
716.65
Special offer
Online exclusive
Ends: 28-Dec-2026
843.00
Save 126.35
Each
Chemical Identifiers
CAS288-32-4
IUPAC Name1H-imidazole
Molecular FormulaC3H4N2
InChI KeyRAXXELZNTBOGNW-UHFFFAOYSA-N
SMILESN1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
pH9.5 to 11.0 (5% at 25°C)
Appearance (Form)Powder or crystals
Titration with HClO4>=99 %
Residue after ignition=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only