Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 20000-30000
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 20000-30000
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 20000-30000
Thermo Scientific Chemicals

Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 20000-30000

CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
1 kg
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 396760010
Price (USD)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS9002-89-5
Molecular Formula(C2H4O)n
SMILESOC(-*)C-*
MDL NumberMFCD00081922
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Almost white to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Degree of hydrolysis87.0 to 89.0 %
Volatile matter=<5.0 % (105°C, 3 h)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only