n-Octane, 99+%, Extra Dry, AcroSeal™
n-Octane, 99+%, Extra Dry, AcroSeal™
n-Octane, 99+%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals

n-Octane, 99+%, Extra Dry, AcroSeal™

CAS: 111-65-9 | C8H18 | 114.23 g/mol
Have Questions?
Catalog NumberQuantity
396901000100 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 396901000
Price (ZAR)
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS111-65-9
IUPAC Nameoctane
Molecular FormulaC8H18
InChI KeyTVMXDCGIABBOFY-UHFFFAOYSA-N
SMILESCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Residue after evaporation=<0.002 %
GC>=99.0 %
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only