1-Propanol, 99.5%, Extra Dry, AcroSeal™
1-Propanol, 99.5%, Extra Dry, AcroSeal™
1-Propanol, 99.5%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals

1-Propanol, 99.5%, Extra Dry, AcroSeal™

CAS: 71-23-8 | C3H8O | 60.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number 396940010
Price (USD)
122.00
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
122.00
Each
Chemical Identifiers
CAS71-23-8
IUPAC Namepropan-1-ol
Molecular FormulaC3H8O
InChI KeyBDERNNFJNOPAEC-UHFFFAOYSA-N
SMILESCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Filtered to 0.2 micrometerFiltered
Water=<0.005 % (Coulometric)
GC>=99.5 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only