5-Formyl-2-thiopheneboronic acid, 97%
5-Formyl-2-thiopheneboronic acid, 97%
5-Formyl-2-thiopheneboronic acid, 97%
Thermo Scientific Chemicals

5-Formyl-2-thiopheneboronic acid, 97%

CAS: 4347-33-5 | C5H5BO3S | 155.96 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3971500101 gGlass bottle
Catalog number 397150010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4347-33-5
IUPAC Name(5-formylthiophen-2-yl)boronic acid
Molecular FormulaC5H5BO3S
InChI KeyDEQOVKFWRPOPQP-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(S1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to orange
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only