Methylcyclohexane, 98+%, Extra Dry, AcroSeal™
Methylcyclohexane, 98+%, Extra Dry, AcroSeal™
Methylcyclohexane, 98+%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals

Methylcyclohexane, 98+%, Extra Dry, AcroSeal™

CAS: 108-87-2 | C7H14 | 98.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number 397240010
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS108-87-2
IUPAC Namemethylcyclohexane
Molecular FormulaC7H14
InChI KeyUAEPNZWRGJTJPN-UHFFFAOYSA-N
SMILESCC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.0 %
Water=<0.005 %
Residue after evaporation=<0.001 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only