5-Fluorosalicylaldehyde, 97%
5-Fluorosalicylaldehyde, 97%
5-Fluorosalicylaldehyde, 97%
Thermo Scientific Chemicals

5-Fluorosalicylaldehyde, 97%

CAS: 347-54-6 | C7H5FO2 | 140.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 397570010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS347-54-6
IUPAC Name5-fluoro-2-hydroxybenzaldehyde
Molecular FormulaC7H5FO2
InChI KeyFDUBQNUDZOGOFE-UHFFFAOYSA-N
SMILESOC1=CC=C(F)C=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or solid
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
Melting point81°C to 86°C
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only