2-Fluoro-5-formylbenzonitrile, 99%
2-Fluoro-5-formylbenzonitrile, 99%
2-Fluoro-5-formylbenzonitrile, 99%
Thermo Scientific Chemicals

2-Fluoro-5-formylbenzonitrile, 99%

CAS: 218301-22-5 | C8H4FNO | 149.12 g/mol
Have Questions?
Catalog NumberQuantity
3979400505 g
Catalog number 397940050
Price (USD)
151.65
Online exclusive
168.00
Save 16.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
151.65
Online exclusive
168.00
Save 16.35
Each
Chemical Identifiers
CAS218301-22-5
IUPAC Name2-fluoro-5-formylbenzonitrile
Molecular FormulaC8H4FNO
InChI KeyMOFRJTLODZILCR-UHFFFAOYSA-N
SMILESFC1=C(C=C(C=O)C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light orange
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
Melting point85°C to 89°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only