3-Ethynylpyridine, 96%
3-Ethynylpyridine, 96%
3-Ethynylpyridine, 96%
Thermo Scientific Chemicals

3-Ethynylpyridine, 96%

CAS: 2510-23-8 | C7H5N | 103.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 398070010
Price (USD)
58.65
Special offer
Online exclusive
Ends: 28-Dec-2026
68.50
Save 9.85
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
58.65
Special offer
Online exclusive
Ends: 28-Dec-2026
68.50
Save 9.85
Each
Chemical Identifiers
CAS2510-23-8
IUPAC Name3-ethynylpyridine
Molecular FormulaC7H5N
InChI KeyCLRPXACRDTXENY-UHFFFAOYSA-N
SMILESC#CC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=95 %
Appearance (Form)Low melting solid
Infrared spectrumConforms
Appearance (Color)Colorless to brown
AppearanceMay darken on storage
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only