N-BOC-p-phenylenediamine, 97%
N-BOC-p-phenylenediamine, 97%
N-BOC-p-phenylenediamine, 97%
Acros Organics

N-BOC-p-phenylenediamine, 97%

CAS: 71026-66-9 | C11H16N2O2 | 208.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 398330050
also known as 39833
HSN 29215990
Price (INR)
25,082.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
25,082.00
Each
Chemical Identifiers
CAS71026-66-9
IUPAC Nametert-butyl N-(4-aminophenyl)carbamate
Molecular FormulaC11H16N2O2
InChI KeyWIVYTYZCVWHWSH-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point113°C to 115°C
HPLC>=96.0 %
Appearance (Color)Beige to brown
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only