2-Hydroxy-3-methylbenzaldehyde, 98%
2-Hydroxy-3-methylbenzaldehyde, 98%
2-Hydroxy-3-methylbenzaldehyde, 98%
Thermo Scientific Chemicals

2-Hydroxy-3-methylbenzaldehyde, 98%

CAS: 824-42-0 | C8H8O2 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 398540010
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.00
Save 5.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.00
Save 5.35
Each
Chemical Identifiers
CAS824-42-0
IUPAC Name2-hydroxy-3-methylbenzaldehyde
Molecular FormulaC8H8O2
InChI KeyIPPQNXSAJZOTJZ-UHFFFAOYSA-N
SMILESCC1=CC=CC(C=O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow to orange
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5620 to 1.5650 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only