5-Bromoindole-2-carboxylic acid, 98%
5-Bromoindole-2-carboxylic acid, 98%
5-Bromoindole-2-carboxylic acid, 98%
Thermo Scientific Chemicals

5-Bromoindole-2-carboxylic acid, 98%

CAS: 7254-19-5 | C9H6BrNO2 | 240.05 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3986500101 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 398650010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7254-19-5
IUPAC Name5-bromo-1H-indole-2-carboxylic acid
Molecular FormulaC9H6BrNO2
InChI KeyYAULOOYNCJDPPU-UHFFFAOYSA-N
SMILESOC(=O)C1=CC2=CC(Br)=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only