5-Bromo-2-chloroanisole, 98%
5-Bromo-2-chloroanisole, 98%
5-Bromo-2-chloroanisole, 98%
Thermo Scientific Chemicals

5-Bromo-2-chloroanisole, 98%

CAS: 16817-43-9 | C7H6BrClO | 221.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 398900050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16817-43-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light brown
Appearance (Form)Liquid or low melting solid
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only