2-Methyl-1-propanol, 99+%, Extra Dry, AcroSeal™
2-Methyl-1-propanol, 99+%, Extra Dry, AcroSeal™
2-Methyl-1-propanol, 99+%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals

2-Methyl-1-propanol, 99+%, Extra Dry, AcroSeal™

CAS: 78-83-1 | C4H10O | 74.12 g/mol
Quantity:
100 mL
1 L
Catalog number 398951000
Price (JPY)
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS78-83-1
IUPAC Name2-methylpropan-1-ol
Molecular FormulaC4H10O
InChI KeyZXEKIIBDNHEJCQ-UHFFFAOYSA-N
SMILESCC(C)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Water=<0.005 %
Residue after evaporation=<0.001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only