(1R,3S)-(+)-4-Cyclopentene-1,3-diol 1-acetate, 99%, stabilized
(1R,3S)-(+)-4-Cyclopentene-1,3-diol 1-acetate, 99%, stabilized
(1R,3S)-(+)-4-Cyclopentene-1,3-diol 1-acetate, 99%, stabilized
Thermo Scientific Chemicals

(1R,3S)-(+)-4-Cyclopentene-1,3-diol 1-acetate, 99%, stabilized

CAS: 60410-16-4 | C7H10O3 | 142.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 399340050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS60410-16-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Solid
Infrared spectrumConforms
Melting point49°C to 53°C
Specific optical rotation+67° to +69° (20°C, 589 nm) (c=2.3, CHCl3)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only