4-Pyridazinecarboxylic acid, 95%
4-Pyridazinecarboxylic acid, 95%
4-Pyridazinecarboxylic acid, 95%
Thermo Scientific Chemicals

4-Pyridazinecarboxylic acid, 95%

CAS: 50681-25-9 | C5H4N2O2 | 124.1 g/mol
Have Questions?
Catalog NumberQuantity
3996300101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 399630010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS50681-25-9
IUPAC Namepyridazine-4-carboxylic acid
Molecular FormulaC5H4N2O2
InChI KeyJUSIWJONLKBPDU-UHFFFAOYSA-N
SMILESOC(=O)C1=CN=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=94 %
Appearance (Color)Off-white to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only