Reusable Cryo Sleeve
Thermo Scientific™
Reusable Cryo Sleeve
Utilize the Thermo Scientific™ Reusable Cryo Sleeve with cane and canister systems.
Have Questions?
Catalog NumberType
4000218Cryo Sleeve
Catalog number 4000218
Price (USD)
197.10
Online exclusive
219.00
Save 21.90
Case of 100
Type:
Cryo Sleeve
Price (USD)
197.10
Online exclusive
219.00
Save 21.90
Case of 100
Utilize the Thermo Scientific™ Reusable Cryo Sleeve with cane and canister systems.
Specifications
DescriptionPolyvinyl Sleeve; I.D. x L: 0.6 in. (1.5cm) x 10.8 in. (27.4cm); Case of 100
For Use With (Equipment)CryoPlus 1,2,3,4
TypeCryo Sleeve
Unit SizeCase of 100