CryoMed™ Sensors
Thermo Scientific™
CryoMed™ Sensors
Improve the functionality of Thermo Scientific™ Cryomed™ controlled-rate freezers with these CryoMed Sensors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
40003864.0/5.0 Cryo Tubes in CryoMed
40003851.2/2.0 Cryo Tubes in CryoMed
40004020.020 Sheath Straw Sample
40004030.062 Sheath Vial Sample
Catalog number 4000386
Price (USD)
-
For Use With (Equipment):
4.0/5.0 Cryo Tubes in CryoMed
Improve the functionality of Thermo Scientific™ Cryomed™ controlled-rate freezers with these CryoMed Sensors.
Features:
  • Thermocouple sensors for cryo tubes, sheath straw and vial samples
  • Includes male plug
Recommended for:
Controlled-rate freezing applications
  • Warranty varies by country, please contact us for details
  • Specifications
    DescriptionThermocouple Sensor
    For Use With (Equipment)4.0/5.0 Cryo Tubes in CryoMed
    HoldsCryo Tube, Sheath Straw, Vial
    Plug TypeMale
    TypeThermocouple Sensor
    Capacity (Metric)5 mL
    Unit SizeEach