Replacement Back-Up Battery for Thermo Scientific™ Ultra-Low Temperature Freezers - FAQs
Replacement Back-Up Battery for Thermo Scientific™ Ultra-Low Temperature Freezers - FAQs
View additional product information for Sensors and Replacement Batteries for Thermo Scientific™ Ultra-Low Temperature Freezers - FAQs (290199, 400159)