Acetylcholine iodide, 99%
Acetylcholine iodide, 99%
Acetylcholine iodide, 99%
Thermo Scientific Chemicals

Acetylcholine iodide, 99%

CAS: 2260-50-6 | C7H16INO2 | 273.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 400180100
Price (USD)
105.00
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
105.00
Each
Chemical Identifiers
CAS2260-50-6
IUPAC Name[2-(acetyloxy)ethyl]trimethylazanium iodide
Molecular FormulaC7H16INO2
InChI KeySMBBQHHYSLHDHF-UHFFFAOYSA-M
SMILES[I-].CC(=O)OCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point156°C to 164°C
Titration Argentometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only