Acridine-9-carboxylic acid hydrate, 97%
Acridine-9-carboxylic acid hydrate, 97%
Acridine-9-carboxylic acid hydrate, 97%
Thermo Scientific Chemicals

Acridine-9-carboxylic acid hydrate, 97%

CAS: 332927-03-4 | C14H9NO2 | 223.23 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 400220250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS332927-03-4
IUPAC Nameacridine-9-carboxylic acid
Molecular FormulaC14H9NO2
InChI KeyIYRYQBAAHMBIFT-UHFFFAOYSA-N
SMILESOC(=O)C1=C2C=CC=CC2=NC2=CC=CC=C12
さらに表示
Water=<15 % (K.F.)
Appearance (Color)Bright yellow to yellow-green
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=96.0 % (on dry substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only