N-Acryloxysuccinimide, 99%
N-Acryloxysuccinimide, 99%
N-Acryloxysuccinimide, 99%
Thermo Scientific Chemicals

N-Acryloxysuccinimide, 99%

CAS: 38862-24-7 | C7H7NO4 | 169.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 400300010
Price (USD)
96.65
Online exclusive
107.00
Save 10.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
96.65
Online exclusive
107.00
Save 10.35
Each
Chemical Identifiers
CAS38862-24-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
Titration with TBAH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only