Agar, pure, powder
Thermo Scientific Chemicals

Agar, pure, powder

For use in biological culture media.
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
1 kg
5 kg
25 kg
Catalog number 400400010
Price (USD)
306.65
Special offer
Online exclusive
Ends: 28-Dec-2026
361.00
Save 54.35
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
306.65
Special offer
Online exclusive
Ends: 28-Dec-2026
361.00
Save 54.35
Each
Chemical Identifiers
CAS9002-18-0
IUPAC Name2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
Molecular FormulaC14H24O9
InChI KeyGYYDPBCUIJTIBM-UHFFFAOYNA-N
SMILESCOC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Ash=<6 %
Appearance (Color)White to cream to light yellow to beige
Appearance (Form)Powder
Infrared spectrumConforms
Loss on drying=<20.0 % (1 g, 105°C, 5 h)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only