Alcian Blue 8GX, certified
Alcian Blue 8GX, certified
Alcian Blue 8GX, certified
Thermo Scientific Chemicals

Alcian Blue 8GX, certified

CAS: 33864-99-2 | C56H68Cl4CuN16S4 | 1298.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 400460100
Price (USD)
175.65
Online exclusive
195.00
Save 19.35
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
175.65
Online exclusive
195.00
Save 19.35
Each
Chemical Identifiers
CAS33864-99-2
IUPAC Name(9Z,18Z,26Z,35Z)-4,14,22,32-tetrakis({[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}methyl)-9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11(16),12,14,17(41),18,20,22,24,26,28(40),29(34),30,32,35-nonadecaene-38,38-bis(ylium) tetrachloride
Molecular FormulaC56H68Cl4CuN16S4
InChI KeyXRWQTDIELZDCPS-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].CN(C)C(SCC1=CC2=C(C=C1)\C1=N\C3=C4C=CC(CSC(N(C)C)=[N+](C)C)=CC4=C4\N=C5/N=C(/N=C6\N([Cu++]N34)\C(=N/C2=N1)C1=CC(CSC(N(C)C)=[N+](C)C)=CC=C61)C1=C5C=C(CSC(N(C)C)=[N+](C)C)C=C1)=[N+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark blue to purple
Infrared spectrumConforms
Dye content>=50 %
UV Lambda max615 to 635 nm (in water)
BSC CertificationPasses
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only