Triethylamine Acetate (TEAA) 2.0 M
Applied Biosystems™

Triethylamine Acetate (TEAA) 2.0 M

This 2.0 M Triethylamine Acetate solution is used with OPC™ cartridges (Cat. No. 400771), for oligonucleotide purification. The Triethylamine AcetateRead more
Have Questions?
Catalog NumberQuantity
400613200 mL
Catalog number 400613
Price (USD)
170.00
Each
Quantity:
200 mL
Price (USD)
170.00
Each
This 2.0 M Triethylamine Acetate solution is used with OPC™ cartridges (Cat. No. 400771), for oligonucleotide purification. The Triethylamine Acetate solution comes in a convenient large volume for ABI 394 Nucleic Acid synthesizers, resulting in extended hands-free DNA synthesis. With these larger bottles attached your synthesizer can produce up to 40 average primer-length oligonucleotides with no bottle changes, reducing attended operation time by 50%.

• Reagent concentrations are within our strict specifications, guaranteeing consistent quality.
• Our performance specification of at least 98% stepwise coupling efficiency is assured, increasing oligonucleotide synthesis yields of full-length product.
• Reagent kits are available for your convenience, and save you time and money.

Our high-performance reagents begin with raw materials that must conform to strict specifications. Once the reagents are manufactured and packaged, we perform extensive testing to assure quality. Reagent lifetime on the instrument is two weeks for tetrazole, and six weeks for all other reagents.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialTriethylamine Acetate
For Use With (Application)Oligo Synthesis
For Use With (Equipment)3900 DNA Synthesizer
FormLiquid
Product TypeOligonucleotide Synthesis Reagent
Quantity200 mL
Reagent TypeSolvent or General Reagent
Sample TypeDNA
Shipping ConditionRoom Temperature
FormatBottle
Unit SizeEach
Contents & Storage
Store in a cool, dry, and well-ventilated location, appropriate for flammable lliquids.

Frequently asked questions (FAQs)

Do you know of any alternatives for Triethylamine Acetate (TEAA) 2.0 M (Cat. No. 400613)?

We do not offer any alternatives for Triethylamine Acetate (TEAA) 2.0 M.