Product Image
Applied Biosystems™

GeneScan™ 1000 ROX™ dye Size Standard

The GeneScan™ 1000 ROX™ Size Standard is an internal lane size standard developed for use with Applied Biosystems™ fluorescence-based DNA electrophoresis systems.
Have Questions?
Catalog NumberQuantity
401098400 Reactions
Catalog number 401098
Price (USD)
448.00
Each
Quantity:
400 Reactions
Price (USD)
448.00
Each

The GeneScan™ 1000 ROX™ Size Standard is an internal lane size standard developed for use with Applied Biosystems™ fluorescence-based DNA electrophoresis systems. The use of an internal lane size standard during electrophoresis enables automated data analysis and precise DNA fragment size comparisons between electrophoresis runs. GeneScan™ 1000 ROX™ Size Standard is designed for sizing DNA fragments in the 47–946 bp range and provides labeled fragments of 47, 51, 55, 82, 85 93 99, 126, 136, 262, 293, 317, 439, 557, 692, 695 and 946 bases. Each of the DNA fragments is labeled with ROX™ fluorophore, which results in a single peak when run under non-denaturing conditions.

Note: Under denaturing conditions using POP-4™ polymer, fragment lengths between 100 and 539 bp can be determined (the linear range of the separation with respect to length vs. scan number).

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Application)Fragment analysis
For Use With (Equipment)3730xl DNA Analyzer, 3730 DNA Analyzer, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer
Label or DyeROX
No. of Reactions400 Reactions
Product LineGeneScan, ROX
Product TypeDNA Seqeuncing Size Standard
Quantity400 Reactions
Shipping ConditionWet Ice
Unit SizeEach
Contents & Storage
This kit contains the size standard.
Store the kit at 2° to 8°C (Do not freeze).

Frequently asked questions (FAQs)

If my fragments are greater than 600 bp, can I use the GeneScan 1200 LIZ Dye, GeneScan 1000 ROX Dye, or GeneScan 2500 ROX Dye Size Standards?

For fragments that are greater than 600 bp, the GeneScan 1200 LIZ Dye Size Standard is recommended. The GeneScan 1000 ROX Dye, or GeneScan 2500 ROX Dye Size Standards are intended for non-denaturing applications only.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.