Product Image
Applied Biosystems™

1- Methylimidazole/THF, Capping Formula B

Cap B is a mix of NMI (N-methylimidazole) and THF (tetrahydrofuran).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
401175450 mL
400785180 mL
43408664 x 180 mL
43408614 x 450 mL
Catalog number 401175
Price (USD)
942.00
Each
Quantity:
450 mL
Price (USD)
942.00
Each

Contains 4 CapB filled Trap-Pak™ bags.

Cap B is a mix of NMI (N-methylimidazole) and THF (tetrahydrofuran).

For Research Use Only. Not for use in diagnostics procedures.

Specifications
For Use With (Equipment)3900 DNA Synthesizer
FormatLiquid
Product Type1-methylimidazole/tetrahydrofuran
Quantity450 mL
Reagent TypeSolvent or General Reagent
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store in a cool, dry, and well-ventilated location, appropriate for flammable lliquids.