Product Image
Applied Biosystems™

Biotin Amidite

Contains 85 mg Biotin Amidite.Biotin Amidite is used for the 5’ attachment of Biotin during oligonucleotide synthesis.The label is treatedRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
401396250 mg
40139585 mg
Catalog number 401396
Price (MXN)
-
Quantity:
250 mg
Contains 85 mg Biotin Amidite.

Biotin Amidite is used for the 5’ attachment of Biotin during oligonucleotide synthesis.

The label is treated as if it were simply another base: It is inserted at the appropriate place in the sequence and incorporated automatically.

• Specialty phosphoramidite allow online addition of the label during synthesis, saving time in production of labeled probes and primers.
• Designed and optimized for use with ABI synthesizers, guaranteeing reliable nucleic acid synthesis.
• We offer a range of specialty phosphoramites and linkers to give you the flexibility demanded in oligonucleotide synthesis.

On-instrument labeling typically saves hours of post-synthesis manual processing.
Biotin Amidite is used to create 5'-biotinylated oligonucleotides that allow control of the oligonucleotide through the affinity of biotin for streptavidin. Capturing biotin-labeled oligonucleotides on streptavidin-coated media simplifies recovery of target sequences and immobilization for sequencing and nonisotopic assays.

Note: See user's manual or package insert for limited label license, and trademark information. For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Oligo Synthesis
For Use With (Equipment)3900 DNA Synthesizer
FormatPowder
Nucleic Acid TypeNone (Modifier)
Nucleoside TypeNone (Modifier)
Product TypeBiotin Amidite
Quantity250 mg
Reagent TypeBiotin Amidite
Sample TypeDNA
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).