Fmoc-L-Valine-HMP Resin - FAQs
Fmoc-L-Valine-HMP Resin - FAQs
View additional product information for Fmoc-L-Valine-HMP Resin - FAQs (401433)
1 product FAQs found
What pore size, loading, and type of resin is recommended for syntheses on the 433A?
The Model 433A Peptide Synthesis System is a batch synthesizer. With Fmoc chemistry, the optimal conditions are: 200-400 "mesh" (or 70-40 micrometer diameter) beads are preferred, but 100-200 mesh (larger beads) will work. This assumes the primary solvent is "NMP" (N-methylpyrrolidone). For the standard chemistries, the loading should be about 0.5 to 0.9 mmole/g (Fmoc protected active group/ per g of resin). Use of lower loaded resins may require both a larger reaction vessel and a different chemistry, or may not be advised. The best resin type is HMP (p-hydroxymethylphenoxymethyl) polystyrene resin, which may also be called "Wang" resin. If PEG (polyethylene glycol-polystyrene) or other "extended" resins are used, adjustment must be made to accommodate the typically lower loading, and less favorable swelling properties.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.