Aluminon, ACS reagent
Aluminon, ACS reagent
Aluminon, ACS reagent
Thermo Scientific Chemicals

Aluminon, ACS reagent

CAS: 569-58-4 | C22H23N3O9 | 473.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 401501000
Price (USD)
122.65
Special offer
Online exclusive
Ends: 28-Dec-2026
144.00
Save 21.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
122.65
Special offer
Online exclusive
Ends: 28-Dec-2026
144.00
Save 21.35
Each
Chemical Identifiers
CAS569-58-4
IUPAC Nametriammonium 5-[(3-carboxylato-4-hydroxyphenyl)(3-carboxylato-4-oxocyclohexa-2,5-dien-1-yl)methyl]-2-hydroxybenzoate
Molecular FormulaC22H25N3O9
InChI KeyUDPQMNAHKKTHHI-UHFFFAOYNA-N
SMILES[NH4+].[NH4+].[NH4+].OC1=CC=C(C=C1C([O-])=O)C(C1C=CC(=O)C(=C1)C([O-])=O)C1=CC=C(O)C(=C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after ignition=<0.2 %
Appearance (Color)Red to yellow-brown to red-brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Sensitivity(to Aluminium) Passes test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

What is the solubility of Aluminon?

Aluminon is soluble in water at a concentration of ~800 g/L at 20°C. It is slightly soluble in ethanol and insoluble in petroleum ether.