GeneScan™ 2500 TAMRA™ dye Size Standard - FAQs
GeneScan™ 2500 TAMRA™ dye Size Standard - FAQs
View additional product information for GeneScan™ 2500 TAMRA™ dye Size Standard - FAQs (401545)
2 product FAQs found
If my fragments are greater than 600 bp, can I use the GeneScan 1200 LIZ Dye, GeneScan 1000 ROX Dye, or GeneScan 2500 ROX Dye Size Standards?
For fragments that are greater than 600 bp, the GeneScan 1200 LIZ Dye Size Standard is recommended. The GeneScan 1000 ROX Dye, or GeneScan 2500 ROX Dye Size Standards are intended for non-denaturing applications only.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
Do you offer any fragment analysis-based kits for non-denaturing applications?
We provide POP Conformational Analysis Polymer (CAP) for non-denaturing polynucleotide fragment analysis applications on the Applied Biosystems 3130/3130xl Genetic Analyzers. However, we do not have a premade kit specifically for this application. To learn more about non-denaturing applications, please see here (http://www.thermofisher.com/us/en/home/life-science/sequencing/fragment-analysis/single-strand-conformation-polymorphism-sscp.html).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.